Learn More
Scientific Industries SI™ Large Sample Set (Model 302)
Product Code. 15527325 Shop All Ohaus Products
Change view
Click to view available options
For Use With (Equipment):
Vortex-Genie 2, Vortex-Genie Pulse
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
15527325 Vortex-Genie 2, Vortex-Genie Pulse Each
1 options
Product Code. 15527325 Supplier Scientific Industries SI™ Supplier No. 83041422
  • ONLINE EXCLUSIVE PRICE
    204.00 €
    164.80 € / Pack of 1
    Save 39.20 €
    19% Off
In Stock
Add to basket
Edge
Add to basket
missing translation for 'validMainframeMsg'
Includes a large array of high-density foam inserts and other accessories for use with the Vortex-Genie 2 and Vortex-Genie Pulse mixers. Scientific Industries SI™ Large Sample Set (Model 302) provides a means for keeping a variety of vessels upright and steady while mixing.
  • Used with the Vortex-Genie 2 family of mixers and the Vortex-Genie Pulse for beakers, flasks and large tubes.
  • Set contains:
    • One Recessed Platform
    • Two Elastic Bands
    • An assortment of Foam Inserts for tubes of various diameters (9mm to 37mm)
    • One Blank Foam Insert provided for user customization
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) Vortex-Genie 2, Vortex-Genie Pulse
Includes 1 Recessed Platform, 2 Elastic Bands, an assortment of Foam Inserts for tubes of various diameters (9mm to 37mm), 1 Blank Foam Insert for customization.