missing translation for 'onlineSavingsMsg'
Learn More

Larotrectinib sulfate, MedChemExpress

Product Code. 16461597
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16461597

Brand: Medchem Express HY12866A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Larotrectinib sulfate (LOXO-101 sulfate; ARRY-470 sulfate) is an ATP-competitive oral, selective inhibitor of the tropomyosin-related kinase (TRK) family receptors, with low nanomolar 50% inhibitory concentrations against all three isoforms (TRKA, B, and C).

Larotrectinib sulfate (LOXO-101 sulfate

Specifications

Chemical Name or Material Larotrectinib sulfate
CAS 1223405-08-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C21H24F2N6O6S
Quantity 50 mg
Synonym LOXO-101 sulfateARRY-470 sulfate
Solubility Information DMSO : 50 mg/mL (94.96 mM; Need ultrasonic) ∣H2O : 2 mg/mL (3.80 mM; ultrasonic and adjust pH to 2 with 1M HCl)
SMILES O=S(O)(O)=O.O=C(N1CC[C@H](O)C1)NC2=C3N=C(N4[C@@H](C5=CC(F)=CC=C5F)CCC4)C=CN3N=C2
Molecular Weight (g/mol) 526.51
Formula Weight 526.51
Percent Purity 98.69%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.