missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LARP1 Rabbit anti-Drosophila, Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13708-1-AP-20UL
This item is not returnable.
View return policy
Description
RNA-binding protein that promotes translation of specific classes of mRNAs downstream of the mTORC1 complex. Associates with the mRNA 5′cap in an MTOR-dependent manner and associates with mRNAs containing a 5′ terminal oligopyrimidine (5′TOP) motif, which is present in mRNAs encoding for ribosomal proteins and several components of the translation machinery. Associates with actively translating ribosomes via interaction with PABPC1/PABP and stimulates translation of mRNAs containing a 5′TOP, thereby regulating cell growth and proliferation. Positively regulates the replication of dengue virus (DENV).Specifications
| LARP1 | |
| Polyclonal | |
| Unconjugated | |
| LARP1 | |
| KIAA0731, La related protein 1, LARP, LARP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23367, 303158, 53567, 73158 | |
| -20°C | |
| Liquid |
| Immunofluorescence, Immunohistochemistry (Paraffin), Immunocytochemistry, Western Blot | |
| 0.51 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6PKG0, Q6ZQ58 | |
| larp, Larp1 | |
| LARP1 Fusion Protein Ag4645 | |
| 20 μL | |
| Primary | |
| Human, Drosophila, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction