missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LASS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 508.00 €
Specifications
| Antigen | LASS1 |
|---|---|
| Dilution | Western Blot : 1-5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30066885
|
Novus Biologicals™
NBP3-30898-25ug |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066808
|
Novus Biologicals™
NBP3-30898-100ug |
100 μg |
508.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LASS1 Polyclonal antibody specifically detects LASS1 in Human samples. It is validated for Western BlotSpecifications
| LASS1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CerS1, LAG1 homolog, ceramide synthase 1, LAG1 longevity assurance homolog 1, LAG1MGC90349, longevity assurance (LAG1, S. cerevisiae) homolog 1, Longevity assurance gene 1 protein homolog 1, Protein UOG-1, UOG1LAG1 longevity assurance homolog 1 (S. cerevisiae), upstream of GDF1 | |
| Synthetic peptide derived from the human LASS1 protein. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| Western Blot : 1-5 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Phosphate buffered saline | |
| 10715 | |
| IgG | |
| Ammonium sulfate precipitation |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title