missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LASS1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30912-100ug
This item is not returnable.
View return policy
Description
LASS1 Polyclonal antibody specifically detects LASS1 in Human samples. It is validated for Western Blot
Specifications
| LASS1 | |
| Polyclonal | |
| Western Blot : 1:400 | |
| CerS1, LAG1 homolog, ceramide synthase 1, LAG1 longevity assurance homolog 1, LAG1MGC90349, longevity assurance (LAG1, S. cerevisiae) homolog 1, Longevity assurance gene 1 protein homolog 1, Protein UOG-1, UOG1LAG1 longevity assurance homolog 1 (S. cerevisiae), upstream of GDF1 | |
| Synthetic peptide derived from the human LASS1 protein. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 10715 | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction