missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LASS3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 508.00 €
Specifications
| Antigen | LASS3 |
|---|---|
| Dilution | Western Blot : 1:400 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|---|---|---|---|---|---|---|---|---|---|
| Code produit | Marque | Quantity | Prix | Quantité et disponibilité | |||||
|
30066775
|
Novus Biologicals™
NBP3-30908-100ug |
100 μg |
508.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30067228
|
Novus Biologicals™
NBP3-30908-25ug |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LASS3 Polyclonal antibody specifically detects LASS3 in Human samples. It is validated for Western BlotSpécification
| LASS3 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CerS3, LAG1 homolog, ceramide synthase 3, LAG1 longevity assurance homolog 3, LAG1 longevity assurance homolog 3 (S. cerevisiae), MGC27091 | |
| Synthetic peptide derived from human LASS3 protein | |
| Antigen Affinity-purified |
| Western Blot : 1:400 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Tris buffered saline, 10mM MgCl2, 10mM ammonium sulfate, and 0.2% lactose | |
| 204219 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit