missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ LASS6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
213.00 € - 508.00 €
Specifications
| Antigen | LASS6 |
|---|---|
| Dilution | Western Blot : 1:400 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30067083
|
Novus Biologicals™
NBP3-30906-100ug |
100 μg |
508.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30066905
|
Novus Biologicals™
NBP3-30906-25ug |
25 μg |
213.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LASS6 Polyclonal antibody specifically detects LASS6 in Human samples. It is validated for Western BlotSpecifications
| LASS6 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CerS6, LAG1 homolog, ceramide synthase 6, LAG1 longevity assurance homolog 6, LAG1 longevity assurance homolog 6 (S. cerevisiae), longevity assurance homolog 6, MGC129949, MGC129950 | |
| Synthetic peptide dervied from human LASS6 protein | |
| Antigen Affinity-purified |
| Western Blot : 1:400 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Tris buffered saline, 10mM MgCl2, 10mM ammonium sulfate, and 0.2% lactose | |
| 253782 | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title