missing translation for 'onlineSavingsMsg'
Learn More

Latanoprost, MedChemExpress

Product Code. 16488528
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16488528 10 mg 10mg
16484397 50 mg 50mg
16483786 25 mg 25mg
16471008 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16488528 Supplier Medchem Express Supplier No. HYB057710mg
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Latanoprost (PHXA41) is a prostaglandin F2α analogue and an agonist for the FP prostanoid receptor, and lowers intraocular-pressure (IOP).

Latanoprost (PHXA41) is a prostaglandin F2α analogue and an agonist for the FP prostanoid receptor, and lowers intraocular-pressure (IOP).

Specifications

Chemical Name or Material Latanoprost
Physical Form Oil
CAS 130209-82-4
Color Colorless
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C26H40O5
Synonym PHXA41
Solubility Information H2O : 100 mg/mL (231.17 mM; Need ultrasonic) ∣Ethanol : 100 mg/mL (231.17 mM; Need ultrasonic) ∣DMSO : ≥ 100 mg/mL (231.17 mM)
SMILES O=C(OC(C)C)CCC/C=C\C[C@@H]1[C@@H](CC[C@@H](O)CCC2=CC=CC=C2)[C@H](O)C[C@@H]1O
Molecular Weight (g/mol) 432.59
Formula Weight 432.59
Percent Purity 99.67%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.