missing translation for 'onlineSavingsMsg'
Learn More

Lauric acid, MedChemExpress

Product Code. 16443469
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16443469 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16443469 Supplier Medchem Express Supplier No. HYY0366
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Lauric acid is a middle chain-free fatty acid with strong bactericidal properties. The EC50s for P. acnes, S.aureus, S. epidermidis, are 2, 6, 4 μg/mL, respectively.

Lauric acid is a middle chain-free fatty acid with strong bactericidal properties. The EC50s for P. acnes, S.aureus, S. epidermidis, are 2, 6, 4 μg/mL, respectively.

Specifications

Chemical Name or Material Lauric acid
Physical Form Solid
CAS 143-07-7
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C12H24O2
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : ≥ 250 mg/mL (1248.00 mM)
SMILES CCCCCCCCCCCC(O)=O
Molecular Weight (g/mol) 200.32
Formula Weight 200.32
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.