missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lauroyl Chloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
POA
Chemical Identifiers
| CAS | 112-16-3 |
|---|---|
| Molecular Formula | C12H23ClO |
| Molecular Weight (g/mol) | 218.76 |
| Synonym | Dodecanoic Acid Chloride, Lauric Acid Chloride, n-Dodecanoyl Chloride |
| IUPAC Name | dodecanoyl chloride |
| SMILES | CCCCCCCCCCCC(=O)Cl |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30470716
|
Toronto Research Chemicals
TRC-L178630-250ML |
250 mL |
POA
250mL |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
30475860
|
Toronto Research Chemicals
TRC-L178630-1L |
1 L |
POA
1 L |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
|
30466892
|
Toronto Research Chemicals
TRC-L178630-50ML |
50 mL |
POA
50mL |
N/A | Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
| 112-16-3 | |
| 218.76 | |
| dodecanoyl chloride |
| C12H23ClO | |
| Dodecanoic Acid Chloride, Lauric Acid Chloride, n-Dodecanoyl Chloride | |
| CCCCCCCCCCCC(=O)Cl |
Specifications
| 112-16-3 | |
| Dodecanoic Acid Chloride, Lauric Acid Chloride, n-Dodecanoyl Chloride | |
| CCCCCCCCCCCC(=O)Cl | |
| 218.76 | |
| Lauroyl Chloride |
| C12H23ClO | |
| InChI=1S/C12H23ClO/c1-2-3-4-5-6-7-8-9-10-11-12(13)14/h2-11H2,1H3 | |
| dodecanoyl chloride | |
| 218.1437 |
Safety and Handling
Recommended Storage : 4°C
Spot an opportunity for improvement?
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.