missing translation for 'onlineSavingsMsg'
Learn More

Lawesson's Reagent, 99%

Product Code. 10124840 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10124840 100 g Glass bottle 100g
10582611 25 g Glass bottle 25g
11328901 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10124840 Supplier Thermo Scientific Acros Supplier No. 210891000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19172-47-5 | C14H14O2P2S4 | 404.452 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For C-S bond formation
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19172-47-5
Molecular Formula C14H14O2P2S4
Molecular Weight (g/mol) 404.452
InChI Key CFHGBZLNZZVTAY-UHFFFAOYSA-N
Synonym lawesson's reagent, lawesson reagent, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane 2,4-disulfide, unii-a4125mq8rx, 1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-, 2,4-disulfide, 2,4-bis 4-methoxyphenyl-2,4-dithioxo-1,3,2,4-dithiadiphosphetane, 2,4-bis 4-methoxyphenyl-1,3,2,4-dithiadiphosphetane-2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulfide, 2,4-bis 4-methoxyphenyl-1,3-dithia-2,4-diphosphetane 2,4-disulphide, 4-methoxyphenylthiophosphoric cyclic di thioanhydride
PubChem CID 87949
IUPAC Name 2,4-bis(4-methoxyphenyl)-2,4-bis(sulfanylidene)-1,3,2$l^{5},4$l^{5}-dithiadiphosphetane
SMILES COC1=CC=C(C=C1)P2(=S)SP(=S)(S2)C3=CC=C(C=C3)OC

Specifications

Color White
Melting Point 228.0°C to 231.0°C
Assay Percent Range 99%
Fieser 10,39; 12,59; 13,38; 15,37; 16,37
Solubility Information Solubility in water: decomposes.
Formula Weight 404.45
Percent Purity 99%
Packaging Glass bottle
Quantity 100 g
Physical Form Powder
Chemical Name or Material Lawesson's Reagent
Show More Show Less
compliance-icons
Product Identifier
  • Lawesson''s Reagent
Signal Word
  • Danger
Hazard Category
  • Substance which\, in contact with water\, emits flammable gases Category 2
Hazard Statement
  • H261-In contact with water releases flammable gases.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P231+P232-Handle under inert gas. Protect from moisture.
  • P335+P334-Brush off loose particles from skin Immerse in cool water/wrap in wet bandages.
  • P314-Get medical advice/attention if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.