missing translation for 'onlineSavingsMsg'
Learn More

BD™ LBS Agar Plates

Product Code. 15796162
Change view
Click to view available options
LBS Agar: 20 x 9mm plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15796162 LBS Agar: 20 x 9mm plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15796162 Supplier BD™ Supplier No. 255011
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

BD™ LBS Agar Plates, also known as Rogosa Agar, is a semi-defined, partially selective medium that is ready to use for the isolation and enumeration of lactobacilli from foods and from intestinal, vaginal, and dental flora.

BD LBS Agar is used for the isolation and enumeration of lactobacilli from foods, dairy products, and human intestinal, vaginal, and dental floras.

  • LBS Agar is more selective in preventing the overgrowth of moulds, streptococci and spreading organisms than tomato juice agar
  • Casein peptone, yeast extract, and ammonium salt provide nitrogen
  • Polysorbate 80 supplies fatty acids necessary for the growth of lactobacilli
  • Manganese and magnesium are growth factors
  • Glucose is a universal energy and carbon source
  • Ammonium citrate, sodium acetate, acetic acid and ferrous sulfate act as inhibitors of streptococci and other contaminating organisms and provide the low pH which is tolerated by lactobacilli but not by many other organisms
  • Phosphate, together with acetate and acetic acid, stabilizes the pH

Supplied in ready to use Stacker™ plates

  • Prepared plated media are ready for immediate use
  • Microbiologically controlled for uncompromised isolation of microorganisms
  • Specialized lid and bottom allow plates to interlock and stack without slipping
  • Specially engineered design easily separates without agitating the sample
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store in dark in original sleeve in 2°-8° C
Detects lactobacilli
Format Prepared culture plates
Product Type Agar Plate
Target Organism Class Lactobacilli
Formulation Per 1 L prepared: Pancreatic Digest of Casein 10 g, Sodium Acetate Hydrate 25 g, Yeast Extract 5 g, Acetic Acid 1.3 mL, Potassium Dihydrogen Phosphate 6 g, Magnesium Sulfate 0.575 g, Ammonium Citrate 2 g, Manganese Sulfate 0.12 g, Glucose 20 g, Ferrous Sulfate 0.034 g, Polysorbate 80 1 g, Agar 15 g
For Use With (Application) isolation and enumeration of lactobacilli from foods and from intestinal, vaginal, and dental flora
Incubation Conditions 35 ± 2° C in an aerobic atmosphere
Incubation Time 48-72 hours
Packaging Type 10 lidded plates per plastic sleeve
Quantity 20 Plates
Diameter (Metric) 90 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.