missing translation for 'onlineSavingsMsg'
Learn More

Lck inhibitor 2, MedChemExpress

Product Code. 16486787
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16486787 5 mg 5mg
16475056 100 mg 100mg
16454088 10 mg 10mg
16430447 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16486787 Supplier Medchem Express Supplier No. HY10644

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lck inhibitor 2 is a bis-anilinopyrimidine inhibitor of tyrosine kinases including LCK, BTK, LYN, SYK, and TXK. The IC50 values are 13nM, 9nM, 3nM, 26nM and 2nM for Lck, Btk, Lyn, Btk and Txk respectively

Lck inhibitor 2 is a bis-anilinopyrimidine inhibitor of tyrosine kinases including LCK, BTK, LYN, SYK, and TXK. The IC50 values are 13nM, 9nM, 3nM, 26nM and 2nM for Lck, Btk, Lyn, Btk and Txk respectively

Specifications

Chemical Name or Material Lck inhibitor 2
CAS 944795-06-6
Color Yellow
Purity Grade Notes Research
Molecular Formula C18H17N5O2
Quantity 5 mg
Solubility Information DMSO : 20 mg/mL (59.64 mM; Need ultrasonic)
SMILES OC1=CC=C(C)C(NC2=CC=NC(NC3=CC(C(N)=O)=CC=C3)=N2)=C1
Molecular Weight (g/mol) 335.36
Formula Weight 335.36
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.