missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LCLAT1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 539.00 €
Specifications
| Antigen | LCLAT1 |
|---|---|
| Dilution | Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18452471
|
Novus Biologicals
NBP1-89204-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18271797
|
Novus Biologicals
NBP1-89204 |
0.1 mL |
539.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LCLAT1 Polyclonal specifically detects LCLAT1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| LCLAT1 | |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 253558 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:LHPRTTGFTFVVDRLREGKNLDAVHDITVAYPHNIPQSEKHLLQGDFPREIHFHVHRYPIDTLPTSKEDLQLWCHKR | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 1-acylglycerol-3-phosphate O-acyltransferase 8, 1-AGPAT 8, Acyl-CoA:lysocardiolipin acyltransferase 1, AGPAT8lysocardiolipin acyltransferase, ALCAT11AGPAT8, EC 2.3.1.-, EC 2.3.1.51, FLJ37965, HSRG1849, LYCAT, lysocardiolipin acyltransferase 1, UNQ1849,1-AGP acyltransferase 8 | |
| LCLAT1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title