missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LDB3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 624.00 €
Specifications
| Antigen | LDB3 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18472111
|
Novus Biologicals
NBP2-14190 |
0.1 mL |
624.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18444221
|
Novus Biologicals
NBP2-14190-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LDB3 Polyclonal antibody specifically detects LDB3 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| LDB3 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CMD1C, CYPHER, KIAA01613, KIAA0613FLJ35865, LDB3Z1, LDB3Z4, LIM domain binding 3, LIM domain-binding protein 3, ORACLE, PDLIM6, PDZ and LIM domain 6, Protein cypher, ZASPLVNC3, Z-band alternatively spliced PDZ-motif protein | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: PIGLYSAETLREMAQMYQMSLRGKASGVGLPGGSLPIKDLAVDSASPVYQAVIKSQNKPEDEADEWARRSSNLQS | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol | |
| 11155 | |
| IgG | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title