missing translation for 'onlineSavingsMsg'
Learn More

Lead(II) cyclohexanebutyrate, 94%, Thermo Scientific™

Product Code. 11473507
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11473507

Brand: Thermo Scientific Alfa Aesar A17280.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 62637-99-4
Molecular Formula C20H34O4Pb
Molecular Weight (g/mol) 545.688
MDL Number MFCD00036403
InChI Key CCJZHCZMNGUOCA-UHFFFAOYSA-L
Synonym lead cyclohexanebutyrate, lead bis 4-cyclohexylbutyrate, cyclohexanebutanoic acid, lead 2+ salt, cyclohexanebutanoic acid, lead 2+ salt 2:1, lead 2+ bis 4-cyclohexylbutanoate, acmc-20akns, lead ii cyclohexanebutyrate, 4-cyclohexylbutanoate; lead 2+, biscyclohexanebutyric acid lead ii salt, $l^ 2-lead 2+ ion bis 4-cyclohexylbutanoate
PubChem CID 112896
IUPAC Name 4-cyclohexylbutanoate;lead(2+)
SMILES C1CCC(CC1)CCCC(=O)[O-].C1CCC(CC1)CCCC(=O)[O-].[Pb+2]

Specifikationer

Quantity 5 g
UN Number UN2291
Formula Weight 545.68
Percent Purity 94%
Chemical Name or Material Lead(II) cyclohexanebutyrate
compliance-icons compliance-icons compliance-icons
Produktbeteckning
  • Lead(II) cyclohexanebutyrate
Signalord
  • Danger
Riskkategorier
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1A
  • Specific target organ toxicity after repeated exposure Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Faroangivelser
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H360Df-May damage the unborn child. Suspected of damaging fertility.
  • H373-May cause damage to organs.
  • H410-Very toxic to aquatic life with long lasting effects.
Skyddsangivelse
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.