missing translation for 'onlineSavingsMsg'
Learn More

Lead in Isooctane standard solution, Specpure™, 0μg/g(0.0g/gal), Thermo Scientific Chemicals

Product Code. 15446915
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446915 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15446915 Supplier Thermo Scientific Alfa Aesar Supplier No. 045354.22
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifikationer

CAS 540-84-1
Odor Nearly odorless
Physical Form Liquid
Molecular Formula C8H18
MDL Number MFCD01094200
UN Number UN1262
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILES CC(C)CC(C)(C)C
IUPAC Name 2,2,4-trimethylpentane
Molecular Weight (g/mol) 114.232
PubChem CID 10907
Quantity 100 g
ChEBI CHEBI:62805
Grade Specpure
Chemical Name or Material Lead in Isooctane standard solution, 0μg/g(0.0g/gal)
Visa mer Visa mindre
compliance-icons compliance-icons compliance-icons
Produktbeteckning
  • Lead in Isooctane standard solution, Specpure, 0µg/g (0.000g/gal)
Signalord
  • Danger
Riskkategorier
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Aspiration toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Faroangivelser
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H304-May be fatal if swallowed and enters airways.
  • H410-Very toxic to aquatic life with long lasting effects.
Skyddsangivelse
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..
Kompletterande information
  • MIXTURE LIST-Contain: Isooctane

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.