missing translation for 'onlineSavingsMsg'
Learn More

Lead in Isooctane standard solution, Specpure™, 1850μg/g(5.0g/gal)

Product Code. 15426905
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15426905 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15426905 Supplier Thermo Scientific Alfa Aesar Supplier No. 045329.22
 Restricted Item
Estimated Shipment: 22-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 540-84-1 | C8H18 | 114.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 540-84-1
Odor Nearly odorless
Physical Form Liquid
Molecular Formula C8H18
MDL Number MFCD01094200
UN Number UN1262
InChI Key NHTMVDHEPJAVLT-UHFFFAOYSA-N
SMILES CC(C)CC(C)(C)C
IUPAC Name 2,2,4-trimethylpentane
Molecular Weight (g/mol) 114.232
PubChem CID 10907
Quantity 100 g
ChEBI CHEBI:62805
Grade Specpure
Chemical Name or Material Lead in Isooctane standard solution, 1850μg/g(5.0g/gal)
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Lead in Isooctane standard solution, Specpure, 1850µg/g (5.0g/gal)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • Aspiration toxicity Category 1
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H304-May be fatal if swallowed and enters airways.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P405-Store locked up.
  • P501-Dispose of contents/container to accordance with local/regional/national/international regulations..
Supplemental information
  • MIXTURE LIST-Contain: Proprietary organo-lead compound, as Pb

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.