missing translation for 'onlineSavingsMsg'
Learn More

Lead Isotopic Standard: Pb @ 100 μg/mL in 2% HNO3 (Natural abundance), VHG™

Product Code. 30003010
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30003010 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30003010 Supplier VHG Labs Supplier No. VHGLISPB150
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Lead Isotopic Standard: Pb @ 100 μg/mL in 2% HNO3 (Natural abundance)
TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material Lead
CAS 7439-92-1
Methods ICP-MS
Analyte or Component Names Lead
Shipping Condition Room Temperature
Molecular Formula Pb
Synonym Plumbum, Lead metal, Blei, Lead, elemental
InChI Formula InChI=1S/Pb
SMILES [Pb]
IUPAC Name lead
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 207.2
Quantity 50 mL
Formula Weight 207.9766 g/mol
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.