missing translation for 'onlineSavingsMsg'
Learn More

Lead Standard, 10,000μg/mL in 2-5% HNO3 (Plasma Grade), SPEX CertiPrep™

Product Code. 15638335 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
500 mL
Packaging:
Triple-leached LDPE bottle
Unit Size:
125mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15638335 500 mL Triple-leached LDPE bottle 500mL
10177733 125 mL Triple-leached LDPE bottle 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15638335 Supplier SPEX Certiprep Supplier No. PLPB23X
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Pb10,000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Chemical Identifiers

CAS 10099-74-8
Molecular Formula N2O6Pb
Molecular Weight (g/mol) 331.20
MDL Number MFCD00011153
InChI Key RLJMLMKIBZAXJO-UHFFFAOYSA-N
PubChem CID 24924
ChEBI CHEBI:37187
SMILES [Pb++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Packaging Triple-leached LDPE bottle
Certificate of Analysis Assurance Certificate of Analysis reports up to 30 elements at ppb levels.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Technique AA, ICP
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Type Certified Reference Material
IUPAC Name lead(2+);dinitrate
Solvent or Matrix 5% HNO3
Quantity 500 mL
Grade Plasma
Concentration or Composition (by Analyte or Components) Matrix Concentration Cat. No.
2-5% HNO3 10, 000μg/mL PLPB2-3Y, -3X
Chemical Name or Material Lead Standard
Afficher plus Afficher moins

Product intended for laboratory use only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.