Learn More
Invitrogen™ Lecanemab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559917
 View more versions of this product
Product Code. 30283188
  • 477.00 € / 100µg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Lecanemab, marketed as Leqembi, is a medication that is prescribed to treat Alzheimer's disease. Lecanemab is a monoclonal antibody derived from a mouse antibody called mAb158, which has been humanized to improve its compatibility with humans. It specifically targets amyloid beta, a protein associated with the disease.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lecanemab Humanized
Recombinant Monoclonal
Unconjugated
BAN2401; Leqembi
Protein A
RUO
Human
Antibody
IgG1 κ
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
100 μg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized