missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LECT1 Monoclonal Antibody (394002)
GREENER_CHOICE

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA561398

Product Code. 30533695

  • 459.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Reconstitute at 0.5 mg/mL in sterile PBS.

ChM-1 is a cartilage-specific matrix glycoprotein that stimulates the growth of chondrocytes. ChM-1 inhibits angiogenesis by disrupting the tube formation of endothelial cells and thus is responsible for the avascular nature of cartilage. ChM-1 is strongly expressed by the proliferating and hypertrophic zones in the epiphyseal plate of long bones. ChM-1 accumulates in the interterritorial matrix around the lacunae. During development, downregulation of ChM-1 permits angiogenesis and ultimately bone formation on the cartilage template. ChM-1 expression is downregulated in the presence of several growth factors including TGFβ2, FGF2 and PTHLH. ChM-1 expression may also play a role in the hypovascularity and chondroid formation of pleomorphic adenomas. The gene encoding human ChM-1 maps to chromosome 13q14-q21.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LECT1
Monoclonal
0.5 mg/mL
PBS with trehalose and no preservative
O75829
CNMD
Mouse myeloma cell line NS0-derived recombinant human Chondromodulin-1 mature chain, Glu215-Val333.
100 μg
Primary
Human
Antibody
IgG2b
Western Blot
394002
Unconjugated
CNMD
BRICD3; BRICHOS domain containing 3; CHM1; Chm-1; Chmi; CHM-I; Chondromodulin; Chondromodulin 1; chondromodulin I; chondromodulin-1; Chondromodulin-I; Chondrosurfactant protein; CH-SP; Cnmd; LECT1; leukocyte cell derived chemotaxin 1; leukocyte cell-derived chemotaxin 1; multiple myeloma tumor suppressor 1; MYETS1
Mouse
Protein A/G
RUO
11061
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Lyophilized
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.