missing translation for 'onlineSavingsMsg'
Learn More

Leflunomide, MedChemExpress

Product Code. 16432006
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16432006 500 mg 500mg
16408086 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16432006 Supplier Medchem Express Supplier No. HYB0083
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Leflunomide is a pyrimidine synthesis inhibitor, inhibiting dihydroorotate dehydrogenase (DHODH), and acts as a disease-modifying antirheumatic drug.

Leflunomide is a pyrimidine synthesis inhibitor, inhibiting dihydroorotate dehydrogenase (DHODH), and acts as a disease-modifying antirheumatic drug.

Specifications

Chemical Name or Material Leflunomide
Physical Form Solid
CAS 75706-12-6
Color White
Quantity 500 mg
Purity Grade Notes Research
Molecular Formula C12H9F3N2O2
For Use With (Application) Cancer-Kinase/protease
Synonym HWA486 RS-34821 SU101
Solubility Information DMSO : ≥ 50 mg/mL (185.04 mM)
SMILES O=C(C1=C(C)ON=C1)NC2=CC=C(C(F)(F)F)C=C2
Molecular Weight (g/mol) 270.21
Formula Weight 270.21
Percent Purity 99.72%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.