missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LEFTY2 Monoclonal Antibody (182515)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561826
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Left-right determination factor 1 and 2 (Lefty1/2) belong to the TGF-beta superfamily of proteins and are responsible for determination of left-right positioning of visceral organs during embryogenesis. Lefty1 acts as a regulator of Lefty2 and NODAL as the lack of the former resulted in bilateral expression of nodal, lefty2 and Pitx2 (a homeobox gene normally expressed on the left side and blocks the signal for leftness). Lefty2 may play a role in endometrial bleeding.
Specifications
| LEFTY2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| O00292 | |
| LEFTY2 | |
| Mouse myeloma cell line NS0-derived recombinant human Lefty-A, Phe78-Pro366. | |
| 500 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 182515 | |
| Unconjugated | |
| LEFTY2 | |
| 6030463A22Rik; AV214969; Ebaf; Ebaf2; endometrial bleeding associated factor; endometrial bleeding-associated factor; LEFTA; Leftb; Left-right determination factor 2; left-right determination factor A; Left-right determination factor B; left-right determination, factor A; LEFTY2; LEFTYA; MGC46222; protein lefty-2; Protein lefty-A; protein lefty-B; PSEC0024; TGFB4; TGF-beta-4; Transforming growth factor beta-4 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 7044 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction