missing translation for 'onlineSavingsMsg'
Learn More

Lepidine, 97%

Product Code. 11477926
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11477926 10 g 10g
11487926 50 g 50g
11497926 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11477926 Supplier Thermo Scientific Alfa Aesar Supplier No. A14040.09
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 491-35-0 | C10H9N | 143.189 g/mol

Lepidine is used in the preparation of certain dyes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lepidine is used in the preparation of certain dyes.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 491-35-0
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.189
MDL Number MFCD00006784
InChI Key MUDSDYNRBDKLGK-UHFFFAOYSA-N
Synonym lepidine, quinoline, 4-methyl, cincholepidine, lepidin, p-methylquinoline, 4-lepidine, gamma-methylquinoline, 4-methyl-quinoline, 4-methyl quinoline, ccris 2894
PubChem CID 10285
ChEBI CHEBI:48983
IUPAC Name 4-methylquinoline
SMILES CC1=CC=NC2=CC=CC=C12

Specifications

Melting Point 9°C to 10°C
Density 1.083
Boiling Point 260°C to 263°C
Flash Point >110°C (230°F)
Refractive Index 1.62
Quantity 10 g
Beilstein 110926
Merck Index 14,5441
Solubility Information Slightly soluble in water.
Formula Weight 143.19
Percent Purity 98%
Chemical Name or Material Lepidine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Lepidine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H341-Suspected of causing genetic defects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Lepidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.