Learn More
Invitrogen™ Leronlimab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA559050
 View more versions of this product
Product Code. 30284566
  • 1204.00 € / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Leronlimab (PRO140) is a humanized IgG4 monoclonal antibody that can target to block chemokine receptor 5 (CCR5), a cell receptor used in HIV infection, tumor metastasis, and immune signal transduction Play multiple roles.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Leronlimab Humanized
Recombinant Monoclonal
Unconjugated
HuPRO-140; PA-14; PRO-140
Protein A
RUO
Human
Antibody
IgG4
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
14.19 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized