missing translation for 'onlineSavingsMsg'
Learn More

Leuco Malachite Green, TRC

Product Code. 30137913
Change view
Click to view available options
Quantity:
100 mg
250 mg
5 g
Unit Size:
100mg
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30137913 5 g 5g
30132757 250 mg 250mg
17992335 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30137913 Supplier Toronto Research Chemicals Supplier No. TRCL3303005G
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129-73-7
Molecular Formula C23 H26 N2
Molecular Weight (g/mol) 330.47
Synonym Benzenamine, 4,4'-(phenylmethylene)bis[N,N-dimethyl-, Aniline, 4,4'-benzylidenebis[N,N-dimethyl- (6CI,7CI,8CI), 4,4'-(Phenylmethylene)bis[N,N-dimethylbenzenamine], 4,4'-Benzylidene bis[N,N-dimethylaniline], 4,4'-Bis(N,N-dimethylaminophenyl)phenylmethane, 4,4'-Bis(dimethylamino)triphenylmethane, Bis(4-dimethylaminophenyl)phenylmethane, Bis(p-dimethylaminophenyl)phenylmethane, Bis[p-(N,N-dimethylamino)phenyl]phenylmethane, C.I. Basic Green 4, leuco base, Leucomalachite green, Malachite green leuco, Malachite green leuco base, NSC 36379, p,p'-Benzylidenebis(N,N-dimethylaniline)
IUPAC Name 4-[[4-(dimethylamino)phenyl]-phenylmethyl]-N,N-dimethylaniline
SMILES CN(C)c1ccc(cc1)C(c2ccccc2)c3ccc(cc3)N(C)C

Specifications

Quantity 5 g
InChI Formula InChI=1S/C23H26N2/c1-24(2)21-14-10-19(11-15-21)23(18-8-6-5-7-9-18)20-12-16-22(17-13-20)25(3)4/h5-17,23H,1-4H3
Formula Weight 330.2096
Percent Purity >95%
Chemical Name or Material Leucomalachite green
Grade HPLC
compliance-icons
Product Identifier
  • Leuco Malachite Green
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Germ cell mutagenicity Category 2
Hazard Statement
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: Leucomalachite Green
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.