missing translation for 'onlineSavingsMsg'
Learn More

Leupeptin hemisulfate, MedChemExpress

Product Code. 16439967
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16439967 5 mg 5mg
16427488 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16439967 Supplier Medchem Express Supplier No. HY18234A5mg
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Leupeptin hemisulfate is a membrane-permeable thiol protease inhibitor that inhibits Cathepsin B, Cathepsin H and Cathepsin L, and also impairs amphisome-lysosome fusion. Leupeptin hemisulfate also exhibits anti-inflammatory effect.

Leupeptin hemisulfate is a membrane-permeable thiol protease inhibitor that inhibits Cathepsin B, Cathepsin H and Cathepsin L, and also impairs amphisome-lysosome fusion. Leupeptin hemisulfate also exhibits anti-inflammatory effect.

Specifications

Chemical Name or Material Leupeptin hemisulfate
Physical Form Powder
CAS 103476-89-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H38N6O4.1/2H2SO4
For Use With (Application) COVID-19-immunoregulation
Solubility Information H2O : 83.33 mg/mL (175.21 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C[C@@H](NC([C@@H](NC([C@@H](NC(C)=O)CC(C)C)=O)CC(C)C)=O)CCCNC(N)=N.[0.5H2SO4]
Molecular Weight (g/mol) 475.59
Formula Weight 475.59
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.