missing translation for 'onlineSavingsMsg'
Learn More

Levofloxacin hydrate, MedChemExpress

Product Code. 16498236
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
5 g
Unit Size:
100mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16498236 100 mg 100mg
16490878 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16498236 Supplier Medchem Express Supplier No. HYB0330A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Levofloxacin hydrate (Levofloxacin hemihydrate) is an orally active antibiotic and is active against both Gram-positive and Gram-negative bacteria. Levofloxacin hydrate inhibits the DNA gyrase and topoisomerase IV. Levofloxacin hydrate can be used for chronic periodontitis, airway inflammation and BK Viremia research.

Levofloxacin hydrate (Levofloxacin hemihydrate) is an orally active antibiotic and is active against both Gram-positive and Gram-negative bacteria. Levofloxacin hydrate inhibits the DNA gyrase and topoisomerase IV. Levofloxacin hydrate can be used for chronic periodontitis, airway inflammation and BK Viremia research.

Specifications

Chemical Name or Material Levofloxacin hydrate
Physical Form Powder
CAS 138199-71-0
Color Off-White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C18H20FN3O4.1/2H2O
For Use With (Application) COVID-19-anti-virus
Synonym Levofloxacin hemihydrate
Solubility Information H2O : ≥ 50 mg/mL (135.00 mM) ∣DMSO : 8.33 mg/mL (22.49 mM; Need ultrasonic)
SMILES O=C(C(C1=O)=CN2[C@@H](C)COC3=C(N4CCN(C)CC4)C(F)=CC1=C23)O.[0.5H2O]
Molecular Weight (g/mol) 370.38
Formula Weight 370.38
Percent Purity 98.76%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.