Learn More
Invitrogen™ Lewis Y Biosimilar Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558692
 View more versions of this product
Product Code. 30283125
  • 1204.00 € / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °. This antibody can also be cross referenced by the term MB 311.
The Lewis Y (Le^y) gene is not actually a single gene but refers to a carbohydrate antigen known as Lewis Y or Le^y antigen, which is part of the Lewis blood group system. This antigen is synthesized by the fucosylation of precursor substances, which involves enzymes encoded by the FUT1 (fucosyltransferase 1) and FUT2 genes. These genes are responsible for the addition of fucose, a hexose deoxy sugar, to the precursor oligosaccharides. The expression of Lewis Y antigen has been extensively studied in the context of cancer biology as it is commonly found on the surface of epithelial and endothelial cells but overexpressed in various types of cancers, including colorectal, gastric, and ovarian carcinomas. The overexpression and altered glycosylation patterns of Le^y antigen in tumors have made it a potential biomarker for cancer diagnosis and a target for immunotherapeutic approaches.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Lewis Y Humanized Biosimilar
Recombinant Monoclonal
Unconjugated
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Protein A
RUO
Human
Antibody
IgG1