missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Lgi1 Antibody (S283-7), Novus Biologicals™
Mouse Monoclonal Antibody
174.00 € - 567.00 €
Specifications
| Antigen | Lgi1 |
|---|---|
| Clone | S283-7 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18666836
|
Novus Biologicals
NBP2-42190 |
0.1 mg |
567.00 €
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18638568
|
Novus Biologicals
NBP2-42190-0.025mg |
0.025 mg |
174.00 €
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Lgi1 Monoclonal antibody specifically detects Lgi1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Lgi1 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Neuroscience | |
| PBS (pH 7.4), 50% Glycerol | |
| epilepsy, partial, EPITEMPIN, Epitempin-1, EPTleucine-rich glioma-inactivated protein 1, ETL1epitempin-1, IB1099, leucine-rich, glioma inactivated 1 | |
| Fusion protein amino acids 37-113 (LRRNT domain and first LRR repeat) of mouse LGI1. Rat: 100% identity (77/77 amino acids identical). Human: 98% identity (76/77 amino acids identical). ∽50% identity with LGI2, LGI3 and LGI4. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| S283-7 | |
| Western Blot 1:1000, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Q9JIA1.1 | |
| 9211 | |
| IgG2a | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title