missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ LHC-9 Medium (1X)

Product Code. 11508866 Shop All Gibco Products
Change view
Click to view available options
Quantity:
500 mL
Unit Size:
500mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11508866 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11508866 Supplier Gibco™ Supplier No. 12680013
 Restricted Item
Estimated Shipment: 03-12-2026
Zum Warenkorb hinzufügen
Edge
Zum Warenkorb hinzufügen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

LHC-9 Medium (1X)

LHC serum-free media are the most widely used products for culture of bronchial epithelial cells. For maximum flexibility and convenience, we offer these products with or without supplementation.

LHC Basal Medium (1X), liquid

Applications:

Formulated for the growth of bronchial epithelial cells with further supplements.

Recommended storage conditions:

2° to 8°C.

Shelf life:

12 months

Intended use(s)

Research use only

LHC-8 Medium and LHC-9 Medium

Applications:

Engineered specifically for the growth of bronchial epithelial cells such as IB3-1, S9, and C38, BEAS-2B, BZR, BBM, Het-1A, NHBE (normal human bronchial epithelial cells), and BBE (bovine bronchial epithelial cells) for asthma, allergy, lung cancer, cystic fibrosis, pulmonary, and esophageal cancer research without further supplements.

Performance and quality testing:

Performance tested in a growth assay using BEAS-2B cells.

Recommended storage conditions:

-5°C to -20°C.

Intended use(s)

Research use only

Reference(s):

Pawliczak R., et. al. (2001) J Biol Chem 276, 44613. Zhao Y.L., et. al. (2000) Carcinogenesis 21, 2005. Liu X., et. al. (1998) Am J Physiol 274, L58. Hei T.K., et. al. (1997) Environ Health Perspect 105, 1085. Lechner J.F., and LaVeck, M.A. (1985) J Tissue Cult Methods 9, 43.

TRUSTED_SUSTAINABILITY

Spezifikation

Cell Line B3-1, S9, C38, BEAS-2B, BZR, BBM, Het-1A, NHBE (normal human bronchial epithelial cells), BBE (bovine bronchial epithelial cells)
Cell Type Epithelial Cells
Classification Serum-free
Concentration 1 X
Form Liquid
Product Type LHC-9 Medium
Serum Level Serum-free
Sterility Sterile-filtered
Sterilization Method Sterile-filtered
With Additives Glutamine, Phenol Red
Quantity 500 mL
Shipping Condition Dry Ice
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Culture Type Mammalian Cell Culture
Mehr anzeigen Weniger anzeigen

For Research Use Only. Not for use in diagnostic procedures.

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.