missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ LHR Antibody (LHCGR/7399), Novus Biologicals™

Mouse Monoclonal Antibody

232.00 € - 462.00 €

Specifications

Antigen LHR
Clone LHCGR/7399
Concentration 0.2 mg/mL
Dilution Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30043702
View Documents
Novus Biologicals™
NBP3-26818-100ug
100 μg
462.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30043789
View Documents
Novus Biologicals™
NBP3-26818-20ug
20 μg
232.00 €
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LHR Monoclonal antibody specifically detects LHR in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications

Specifications

LHR
0.2 mg/mL
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Caveolae Markers, GPCR
10 mM PBS with 0.05% BSA
3973
IgG2 κ
Protein A or G purified
LHCGR/7399
Immunohistochemistry-Paraffin 1-2 μg/mL for 30 minutes at RT
Monoclonal
Purified
RUO
Human
FLJ41504, HHG, hypergonadotropic hypogonadism, LCGRLH/CGR, LH/CG-R, LHRHR, LHRLGR2ULG5, LSH-R, Luteinizing hormone receptor, luteinizing hormone/choriogonadotropin receptor, lutropin/choriogonadotropin receptor, lutropin-choriogonadotropic hormone receptor
Recombinant fragment (around aa 100-300) of human LHR protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.