missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LHX9 Antibody (CL15033) [Unconjugated], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-43644-100ul
This item is not returnable.
View return policy
Description
LHX9 Monoclonal antibody specifically detects LHX9 in Human samples. It is validated for Western Blot
Specifications
| LHX9 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1 μg/mL | |
| LIM homeobox 9, LIM homeobox protein 9, LIM/homeobox protein Lhx9 | |
| This antibody was generated using a synthetic peptide of Q9NQ69, with the exact immunogen sequence remaining proprietary. | |
| 100 μL | |
| Cell Biology | |
| 56956 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
| Western Blot | |
| CL15033 | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction