missing translation for 'onlineSavingsMsg'
Learn More

Gilac™ Lid for Dough Container

Product Code. 17988941
Change view
Click to view available options
For Use With (Equipment):
Dough Container
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) Unit Size
17988941 Dough Container Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17988941 Supplier Gilac™ Supplier No. G179221
  • ONLINE EXCLUSIVE PRICE
    16.55 €
    10.89 € / Pack of 1
    Save 5.66 €
    34% Off
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Lid for Dough Container providing effective protection for sensitive contents and/or the last container on the pile.

  • Lid providing effective protection for sensitive contents and/or the last container on the pile.

Specifications

Product Type Lid
Material Polyethylene
Color White
Dimensions (L x W x H) 600 x 400 x 25 mm
For Use With (Equipment) Dough Container
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.