missing translation for 'onlineSavingsMsg'
Learn More

Ratiolab™ Lids for Micro Test Plates

Product Code. 15728148
Change view
Click to view available options
Plate Compatibility:
96-well Micro Test Plate
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Plate Compatibility unitSize
15728148 96-well Micro Test Plate Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15728148 Supplier Ratiolab™ Supplier No. 6018402

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

For use with 96-well micro test plates.

  • Manufactured from clear polystyrene
  • Provides secure closure and protection
  • Provided in packs of 100

Specifications

Color Clear
Material Polystyrene
No. per Pack 100
Product Type Lid
Plate Compatibility 96-well Micro Test Plate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.