missing translation for 'onlineSavingsMsg'
Learn More

Lifirafenib, MedChemExpress

Product Code. 16497556
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497556

Brand: Medchem Express HY18957

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lifirafenib (BGB-283) is a novel and potent Raf Kinase and EGFR inhibitor with IC50 values of 23 and 29 nM for recombinant BRafV600E and EGFR, respectively.

Lifirafenib (BGB-283) is a novel and potent Raf Kinase and EGFR inhibitor with IC50 values of 23 and 29 nM for recombinant BRafV600E and EGFR, respectively.

Specifications

Chemical Name or Material Lifirafenib
CAS 1446090-79-4
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H17F3N4O3
Quantity 100 mg
Synonym BGB-283
Solubility Information DMSO : ≥ 100 mg/mL (209.02 mM)
SMILES O=C1NC2=NC=CC(OC3=CC=C(O[C@@]4([H])[C@]5([H])[C@@H]4C6=NC7=CC=C(C(F)(F)F)C=C7N6)C5=C3)=C2CC1
Molecular Weight (g/mol) 478.42
Formula Weight 478.42
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.