missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIFR alpha Rabbit anti-Mouse, Clone: 110, Novus Biologicals™
Rabbit Monoclonal Antibody
367.00 € - 552.00 €
Specifications
| Antigen | LIFR alpha |
|---|---|
| Clone | 110 |
| Dilution | ELISA 1:5000-1:10000 |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18243247
|
Novus Biologicals
NBP2-90580-50UL |
50 μg |
367.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18274576
|
Novus Biologicals
NBP2-90580-100UL |
100 μg |
552.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIFR alpha Monoclonal specifically detects LIFR alpha in Mouse samples. It is validated for ELISA.Specifications
| LIFR alpha | |
| ELISA 1:5000-1:10000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| 0.2 um filtered solution in PBS | |
| 3977 | |
| IgG | |
| Protein A purified |
| 110 | |
| ELISA | |
| Unconjugated | |
| Rabbit | |
| Mouse | |
| CD118, CD118 antigen, FLJ98106, FLJ99923, leukemia inhibitory factor receptor, leukemia inhibitory factor receptor alpha, LIF receptor, LIF-R, SJS2, STWS, SWS | |
| Recombinant Mouse LIFR alpha protein | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title