missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Alkaline Phosphatase Antibody Labeling Kit

Product Code. 18038494 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 mg
100 μg
30 μg
300 μg
Unit Size:
1 set
100µg
1mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18038494 100 μg 100µg
18700404 300 μg 1 set
18738793 1 mg 1mg
18716733 30 μg 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 18038494 Supplier Novus Biologicals™ Supplier No. 7020005

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applications: ELISA

Alkaline phosphatase (AP) is a hydrolyase enzyme that is frequently conjugated to antibodies for use in immunoassays. PNPP is a popular colorimetric substrate for AP; the product of the reaction, p-nitrophenol, can be read at 405nM. Fluorogenic substrates (e.g. 4-methylumbelliferyl phosphate; MUP) may also be employed.

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 100 μg
Conjugate Alkaline Phosphatase
Kit Contents Glass vial(s) of Lightning-LinkTM mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent
Product Type Alkaline Phosphatase Antibody Labeling Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.