missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Fluorescein Antibody Labeling Kit

Product Code. p-7070041 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
30 μg
300 μg
Unit Size:
1 set
1mg
This item is not returnable. View return policy

Product Code. 18795113

Brand: Novus Biologicals™ 7070030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applications: Immunofluorescence

Fluorescein is a fluorophore widely used in biological analysis, with a pH dependent absorption and emission spectrum. In TBS LL-Fluorescein shows an absorption maximum at 494 nm and an emission maximum at 520 nm. If your antibody needs to be concentrated, purified, or cleaned up, consider purchasing an antibody clean up kit.

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 30 μg
Conjugate FITC
Kit Contents Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent
Product Type Fluorescein Antibody Labeling Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.