missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) HRP Antibody Labeling Kit

Product Code. 18715773 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
1 x 100 μg Reaction
3 x 10 μg Reaction
3 x 100 μg Reaction
5 mg
5 x 1 mg Reaction
Unit Size:
1 set
100µg
1mg
300µg
5mg
This item is not returnable. View return policy

Product Code. 18715773

Brand: Novus Biologicals™ 7010030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Applications: Western Blot, Immunohistochemistry, In vivo assay

Horseradish peroxidase is a 44kDa glycoprotein with 6 lysine residues which can be conjugated to labeled biomolecules using Lightning-Link. The enzyme label can be visualized by chromogenic reactions; for example diaminobenzidine (DAB) in the presence of hydrogen peroxide (H202) is converted in to a water insoluble brown pigment. Other substrates which can be used to measure horseradish peroxidase activity include ABTS, TMB and TMBUS.

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 3 x 10 μg Reaction
Conjugate HRP
Kit Contents Glass vial(s) of Lightning-Link mix (1, 3 or 5 vials, depending on pack size), 1 vial of LL Modifier reagent, 1 vial of LL Quencher reagent
Product Type HRP Antibody Labeling Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.