missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Horseradish peroxidase is a 44kDa glycoprotein with 6 lysine residues which can be conjugated to labeled biomolecules using Lightning-Link. The enzyme label can be visualized by chromogenic reactions; for example diaminobenzidine (DAB) in the presence of hydrogen peroxide (H202) is converted in to a water insoluble brown pigment. Other substrates which can be used to measure horseradish peroxidase activity include ABTS, TMB and TMBUS.
Order Info
Quantity
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 3 x 100 μg Reaction |
| Conjugate | HRP |
| Kit Contents | Glass vial(s) of Lightning-Link mix (1 or 3 vials, depending on pack size), 1 vial of LL-AP Modifier reagent, 1 vial of LL-AP Quencher reagent |
| For Use With (Application) | In vivo assay |
| Product Type | HRP Antibody Labeling Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?