missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Lightning-Link (R) Rapid Cy3 Antibody Labeling Kit

Product Code. 18788693 Shop All Bio Techne Products
Click to view available options
Quantity:
1 mg
30 μg
300 μg
Unit Size:
1 set
1mg
This item is not returnable. View return policy

Product Code. 18788693

Brand: Novus Biologicals™ 3400030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody labeling kit

Cyanine Dye 3 (Cy3) has a strong absorption peak at 550nm and a secondary absorption peak at 520nm. The maximum emission peak is at 570nm, with an excitation coefficient of 1.5 x10&superscript_begin;5&superscript_end; cm&superscript_begin;-1&superscript_end;M&superscript_begin;-1&superscript_end;. Cy3 can be used as an alternative to Alexa Fluor 555, DY-550, Rhodamine Red-X and TAMRA [5(6)-Carboxytetramethylrhodamine].

Order Info

Quantity

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Quantity 30 μg
Conjugate Cy3
Kit Contents Glass vial(s) of Lightning-Link Rapid mix (1 or 3 vials, depending on pack size), 1 vial of Rapid Modifier reagent, 1 vial of Rapid Quencher reagent
Product Type Rapid Cy3 Antibody Labeling Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.