missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Lightning-Link (R) Rapid Fluorescein Antibody Labeling Kit
Shop All Bio Techne ProductsDescription
Fluorescein is a fluorophore widely used in biological analysis, with a pH dependent absorption and emission spectrum. In water it shows an absorption maximum at 494 nm and an emission maximum of 521 nm.
Order Info
Quantity
Specifications
Specifications
| Content And Storage | Store at -20°C. Avoid freeze-thaw cycles. |
| Quantity | 30 μg |
| Conjugate | Fluorescein |
| Product Type | Rapid Fluorescein Antibody Labeling Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?