missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Limitin Monoclonal Antibody (183707)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561609
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Has antiviral and antiproliferative activities. Produced by macrophages and stimulates the production of two enzymes: a protein kinase and an oligoadenylate synthetase.
Specifications
| Limitin | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q61716 | |
| Ifna11 | |
| Mouse myeloma cell line NS0-derived recombinant mouse Limitin/IFN-zeta , Leu22-Arg182. | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| ELISA, Neutralization | |
| 183707 | |
| Unconjugated | |
| Ifna11 | |
| 6030405N23Rik; BGIF; IFN-[a]11; Ifna11; IFN-alpha-11; Ifnz; IFN-z; interferon alpha 11; interferon alpha family, gene 11; interferon alpha-11; interferon zeta; Limitin; Lmtn | |
| Rat | |
| Protein A/G | |
| RUO | |
| 15964 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction