missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIMPII/SR-B2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 17 publications
227.00 € - 587.00 €
Specifications
| Antigen | LIMPII/SR-B2 |
|---|---|
| Dilution | Western Blot 1:1000, Simple Western, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin reported in scientific literature (PMID 23863627) |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18748733
|
Novus Biologicals
NB400-129SS |
0.025 mL |
227.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18735213
|
Novus Biologicals
NB400-129 |
0.1 mL |
587.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LIMPII/SR-B2 Polyclonal specifically detects LIMPII/SR-B2 in Human, Mouse, Rat, Canine, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.Specifications
| LIMPII/SR-B2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cellular Markers, Cholesterol Metabolism, Lipid and Metabolism, Lysosome Markers, Signal Transduction | |
| Whole antisera with 0.05% Sodium Azide | |
| 950 | |
| A C-terminal synthetic peptide made to the mouse LIMPII/lgp85 protein sequence. [UniProt# O35114] | |
| Primary | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot 1:1000, Simple Western, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 1:50-1:250, Immunoprecipitation 1:100, Immunohistochemistry-Paraffin reported in scientific literature (PMID 23863627) | |
| Polyclonal | |
| Antisera | |
| RUO | |
| Human, Mouse, Rat, Canine, Primate | |
| AMRF, CD36 antigen, CD36 antigen (collagen type I receptor, thrombospondin receptor)-like 2(lysosomal integral membrane protein II), CD36L2LIMP II, HLGP85, LGP85, LIMP-2,85 kDa lysosomal sialoglycoprotein scavenger receptor class B, member 2, LIMPIICD36 antigen-like 2, lysosome membrane protein 2, Lysosome membrane protein II, Scavenger receptor class B member 2,85 kDa lysosomal membrane sialoglycoprotein, scavenger receptor class B, member 2, SR-BII | |
| SCARB2 | |
| IgG | |
| Unpurified | |
| 85 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title