missing translation for 'onlineSavingsMsg'
Learn More

LIMPII/SR-B2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 12 publications

220.00 € - 533.00 €

Specifications

Form Antisera
Isotype IgG
View More Specs

Products 2
Product Code Brand dimQuantity Price Quantity & Availability  
Product Code Brand dimQuantity Price Quantity & Availability  
18766643
View Documents
Novus Biologicals
NB400-102SS
220.00 €
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18726313
View Documents
Novus Biologicals
NB400-102
533.00 €
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

LIMPII/SR-B2 Polyclonal specifically detects LIMPII/SR-B2 in Human, Mouse, Rat, Bovine, Chinese Hamster, Hamster, Mustelid samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications

Specifications

Antisera
IgG
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.