missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LIN7B Monoclonal Antibody (OTI1G1), TrueMAB™, OriGene
Description
LIN7B Monoclonal antibody specifically detects LIN7B in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot
Specifications
Specifications
| Antigen | LIN7B |
| Applications | Flow Cytometry, Immunocytochemistry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI1G1 |
| Concentration | 0.39 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | EFNA2 |
| Gene Accession No. | O43921 |
| Gene Alias | hLin7B; hVeli2; lin 7 homolog b; lin-7 homolog B; lin-7 homolog B (C. elegans); lin-7 homolog B, crumbs cell polarity complex component; lin-7-A; LIN7B; lin-7B; MALS2; MALS-2; Mammalian lin-seven protein 2; protein lin-7 homolog B; UNQ3116/PRO10200; Veli1a; Veli2; Veli-2; veli-2 protein; vertebrate homolog of C. elegans Lin-7 type 2; vertebrate lin-7 homolog 2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?