missing translation for 'onlineSavingsMsg'
Learn More

LIN9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Product Code. 16837884 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16837884 20 μL 20µL
16827884 150 μL 150µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16837884 Supplier Proteintech Supplier No. 178821AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

LIN9 belongs to the lin-9 family and acts as a tumor suppressor. It inhibits DNA synthesis. Its ability to inhibit oncogenic transformation is mediated through its association with RB1. LIN9 plays a role in the expression of genes required for the G1/S transition.

Specifications

Antigen LIN9
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 0.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene LIN9
Gene Accession No. Q5TKA1, Q8C735
Gene Alias BARA, BARPsv, hLin 9, HuLin 9, Lin 9, lin 9 homolog (C. elegans), LIN9, pRB associated protein, Protein lin 9 homolog, TGS, TGS1, TUDOR gene similar protein
Gene Symbols LIN9
Host Species Rabbit
Immunogen LIN9 Fusion Protein Ag11889
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 286826, 689523, 72568
Target Species Rat, Mouse, Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.