missing translation for 'onlineSavingsMsg'
Learn More

Linalool 1000 μg/mL in Isopropanol, Dr. Ehrenstorfer

Product Code. 30318723
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30318723 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30318723 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010048IP
 Restricted Item
Estimated Shipment: 31-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material DL-Linalool
CAS 78-70-6
Analyte or Component Names DL-Linalool
Shipping Condition Room Temperature
Molecular Formula C10 H18 O
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym 1,6-Octadien-3-ol, 3,7-dimethyl-, 2,6-Dimethyl-2,7-octadien-6-ol, 2-Methyl-1-prenyl-3-buten-2-ol, 3,7-Dimethyl-1,6-octadien-3-ol, 3,7-Dimethyl-1,6-octadiene-3-ol, 3,7-Dimethyl-3-hydroxy-1,6-octadiene, 3,7-Dimethyl-3-ol-1,6-octadiene, L 260-2, Linalol, Linalool, Linalyl alcohol, Linanool, NSC 3789, Phantol, dl-Linalool, (+/-)-Linalool, beta-Linalool
InChI Formula InChI=1S/C10H18O/c1-5-10(4,11)8-6-7-9(2)3/h5,7,11H,1,6,8H2,2-4H3
SMILES CC(=CCCC(C)(O)C=C)C
IUPAC Name 3,7-dimethylocta-1,6-dien-3-ol
Solvent or Matrix Isopropanol
Molecular Weight (g/mol) 154.249
Quantity 1 mL
Formula Weight 154.136 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.